Tetrapentylammonium bromide, 99+%, Thermo Scientific Chemicals
Tetrapentylammonium bromide, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrapentylammonium bromide, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 221481000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS866-97-7
IUPAC Nametetrapentylazanium bromide
Molecular FormulaC20H44BrN
InChI KeySPALIFXDWQTXKS-UHFFFAOYSA-M
SMILES[Br-].CCCCC[N+](CCCCC)(CCCCC)CCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Melting point99°C to 103°C
Titration Argentometric>=99.0 %
Appearance (Form)Powder or crystals or flakes
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only