Thermo Scientific Chemicals

N-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide, 98%, Thermo Scientific Chemicals

Catalog number: 221540010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

N-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide, 98%, Thermo Scientific Chemicals

Catalog number: 221540010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 221540010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
77377-52-7
IUPAC Name
N-(butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide
Molecular Formula
C9H18F3NOSi
InChI Key
NEJIKZHUEMMAEF-UHFFFAOYSA-N
SMILES
CCCC[Si](C)(C)N(C)C(=O)C(F)(F)F
GC
>=97.5 %
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.4010 to 1.4030 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent, Silylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text