N-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide, 98%, Thermo Scientific Chemicals
N-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 221540250
Price (USD)/ Each
319.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
376.00 
Save 56.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
319.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
376.00 
Save 56.35 (15%)
Add to cart
N-(tert-Butyldimethylsilyl)-N-methyl-trifluoroacetamide, 98%, Thermo Scientific Chemicals
Catalog number221540250
Price (USD)/ Each
319.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
376.00 
Save 56.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC NameN-(butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide
Molecular FormulaC9H18F3NOSi
InChI KeyNEJIKZHUEMMAEF-UHFFFAOYSA-N
SMILESCCCC[Si](C)(C)N(C)C(=O)C(F)(F)F
Molecular Weight (g/mol)241.33
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4010 to 1.4030 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent, Silylation reagent
RUO – Research Use Only