Thermo Scientific Chemicals

Allyl chloroformate, 97%, Thermo Scientific Chemicals

Catalog number: 221740500
50 mL, Glass bottle, Each
Thermo Scientific Chemicals

Allyl chloroformate, 97%, Thermo Scientific Chemicals

Catalog number: 221740500
50 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 221740500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2937-50-0
IUPAC Name
prop-2-en-1-yl carbonochloridate
Molecular Formula
C4H5ClO2
InChI Key
CAEWJEXPFKNBQL-UHFFFAOYSA-N
SMILES
ClC(=O)OCC=C
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.4210 to 1.4240 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text