Allyl chloroformate, 97%, Thermo Scientific Chemicals
Allyl chloroformate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Allyl chloroformate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
50 mL
Packaging:
Glass bottle
Catalog number 221741000
Price (USD)/ Each
222.65
Online exclusive
247.00 
Save 24.35 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
222.65
Online exclusive
247.00 
Save 24.35 (10%)
Add to cart
Allyl chloroformate, 97%, Thermo Scientific Chemicals
Catalog number221741000
Price (USD)/ Each
222.65
Online exclusive
247.00 
Save 24.35 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Nameprop-2-en-1-yl carbonochloridate
Molecular FormulaC4H5ClO2
InChI KeyCAEWJEXPFKNBQL-UHFFFAOYSA-N
SMILESClC(=O)OCC=C
Molecular Weight (g/mol)120.53
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4210 to 1.4240 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only