Thermo Scientific Chemicals

Phenol, 99.5%, extra pure, loose crystals, unstabilized, Thermo Scientific Chemicals

Catalog number: 221755000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Phenol, 99.5%, extra pure, loose crystals, unstabilized, Thermo Scientific Chemicals

Catalog number: 221755000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 221755000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108-95-2
IUPAC Name
phenol
Molecular Formula
C6H6O
InChI Key
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1
Infrared spectrum
Conforms
Water
=<0.2 %
Appearance (Color)
Colorless to white
GC
>=99.5 %
Acidity
(per Ph. Eur.) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text