2-Chloroethyl methyl ether, 98%, Thermo Scientific Chemicals
2-Chloroethyl methyl ether, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloroethyl methyl ether, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Catalog number 221771000
Price (USD)/ Each
69.90
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
69.90
Add to cart
2-Chloroethyl methyl ether, 98%, Thermo Scientific Chemicals
Catalog number221771000
Price (USD)/ Each
69.90
-
Add to cart
Chemical Identifiers
CAS627-42-9
IUPAC Name1-chloro-2-methoxyethane
Molecular FormulaC3H7ClO
InChI KeyXTIGGAHUZJWQMD-UHFFFAOYSA-N
SMILESCOCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.4070 to 1.4100 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only