Thermo Scientific Chemicals

Methyl 3-bromopropionate, 97%, Thermo Scientific Chemicals

Catalog number: 221790250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 3-bromopropionate, 97%, Thermo Scientific Chemicals

Catalog number: 221790250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 221790250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3395-91-3
IUPAC Name
methyl 3-bromopropanoate
Molecular Formula
C4H7BrO2
InChI Key
KQEVIFKPZOGBMZ-UHFFFAOYSA-N
SMILES
COC(=O)CCBr
Appearance (Form)
Liquid
GC
>=96.0 %
Refractive index
1.4565 to 1.4595 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text