Methyl 3-bromopropionate, 97%, Thermo Scientific Chemicals
Methyl 3-bromopropionate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3-bromopropionate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 221790500
Price (USD)/ Each
170.00
-
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
170.00
Add to cart
Methyl 3-bromopropionate, 97%, Thermo Scientific Chemicals
Catalog number221790500
Price (USD)/ Each
170.00
-
Add to cart
Chemical Identifiers
CAS3395-91-3
IUPAC Namemethyl 3-bromopropanoate
Molecular FormulaC4H7BrO2
InChI KeyKQEVIFKPZOGBMZ-UHFFFAOYSA-N
SMILESCOC(=O)CCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4565 to 1.4595 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only