Thermo Scientific Chemicals

Benzoic acid, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 221802500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Benzoic acid, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog number: 221802500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 221802500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
65-85-0
IUPAC Name
benzoic acid
Molecular Formula
C7H6O2
InChI Key
WPYMKLBDIGXBTP-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1
Appearance (Color)
White
Melting point
121°C to 123°C
Sulfated ash
=<0.01 %
Iron (Fe)
=<4 ppm
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text