Appearance (Color)Clear colorless to slightly colored
Water=<0.1 % (K.F.)
Specific gravity(20°C) 1.385 to 1.415
Appearance (Form)Liquid
Refractive index1.4750 to 1.4790 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.