Search Thermo Fisher Scientific
Extend the capabilities and performance of your Thermo Scientific™ Dionex™ Inuvion™ IC system with a system upgrade. Add electrolytic suppression and eluent degas to your Thermo Scientific™ Dionex™ Inuvion™ Core IC system to convert it to a full Dionex Inuvion IC system. You can also upgrade your Dionex Inuvion IC system by adding RFIC™ eluent generation to take advantage of the full benefits of reagent-free ion chromatography.
Type | Upgrade Kit |
Description | Dionex Inuvion IC System to Dionex Inuvion IC System with RFIC Upgrade |
Unit Size | Each |
Type | Upgrade Kit |
Description | Dionex Inuvion Core IC System to Dionex Inuvion IC System Upgrade |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
22185-62002 | Each | Dionex Inuvion IC System to Dionex Inuvion IC System with RFIC Upgrade | 10,450.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
22185-62001 | Each | Dionex Inuvion Core IC System to Dionex Inuvion IC System Upgrade | 7,150.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Note: This option is not available for the Dionex Inuvion Core IC system unless it has already been upgraded to the Dionex Inuvion IC system.