Search Thermo Fisher Scientific
Control your separation temperature with confidence using the Thermo Scientific™ Dionex™ Inuvion™ Column Heater. Featuring active eluent pre-heating and still air thermostatting, it offers reliable and reproducible IC performance and simple method transfer from older instruments.
For Use With (Equipment) | Dionex Inuvion IC System |
Temperature Range | 10 to 60 °C (50 to 140 °F); minimum working range 5 °C above ambient |
Description | Dionex Inuvion Column Heater |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
22185-62400 | Each | Dionex Inuvion Column Heater | 1,500.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|