Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Get low-pulsation regenerant and reagent delivery with the Thermo Scientific™ Dionex™ Inuvion™ integrated peristaltic pump, which neatly integrates into the Thermo Scientific™ Dionex™ Inuvion™ IC system.
For Use With (Equipment) | Dionex Inuvion IC System |
Min. Flow Rate | 0.004 mL/min |
Max. Flow Rate | 315 mL/min. |
Description | Dionex Inuvion Integrated Regenerant Pump |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
22185-62702 | Each | Dionex Inuvion Integrated Regenerant Pump | 1,000.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|