Search Thermo Fisher Scientific
Catalog Number | Description | Type | Pressure |
---|---|---|---|
22185-62703 | Two-Position High Pressure Auxiliary Valve (10 port) for Dionex Inuvion IC System | High-Pressure Valve | 35 MPa (5000 psi) |
22185-62704 | Two-Position High Pressure Auxiliary Valve (6 port) for Dionex Inuvion IC System | High-Pressure Valve | 35 MPa (5000 psi) |
B51001290 | Low-Pressure Auxiliary Valve for Dionex Inuvion IC System | Low-Pressure Valve Assembly | 0.7 MPa (100 psi) |
Extend the application capability of your Thermo Scientific™ Dionex™ Inuvion™ IC system with auxiliary valves. The Dionex Inuvion IC system can support up to two additional low-pressure valves or one additional 6-port or 10-port high-pressure valve.
Share catalog number, name or link