Dionex™ Inuvion™ Digital Gas Pressure Regulator
Dionex™ Inuvion™ Digital Gas Pressure Regulator
Thermo Scientific™
Dionex™ Inuvion™ Digital Gas Pressure Regulator
Maximize day-to-day run consistency with the integrated Thermo Scientific™ Dionex™ Inuvion™ Digital Gas Pressure Regulator.
Have Questions?
Catalog NumberDescriptionFor Use With (Equipment)
22185-62706Dionex Inuvion Digital Gas Pressure RegulatorDionex Inuvion IC System
Catalog number 22185-62706
Price (USD)
1,120.00
Each
-
Add to cart
Description:
Dionex Inuvion Digital Gas Pressure Regulator
For Use With (Equipment):
Dionex Inuvion IC System
Price (USD)
1,120.00
Each
Add to cart
Ask our AI about this Product
Maximize day-to-day run consistency with the Thermo Scientific™ Dionex™ Inuvion™ Digital Gas Pressure Regulator. This optional accessory integrates gas regulation into the Dionex Inuvion IC system with high-visibility, simplified Thermo Scientific™ Chromeleon™ Chromatography Data System (CDS) control, and traceability.
Optional accessory for all Dionex Inuvion IC system configurations.
Specifications
DescriptionDionex Inuvion Digital Gas Pressure Regulator
Flow Rate28 dm3/min (17 SCFM)
For Use With (Equipment)Dionex Inuvion IC System
Pressure0.2 MPa (30 psi)
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link