Dionex™ Eluent Monitor
Dionex™ Eluent Monitor
Thermo Scientific™
Dionex™ Eluent Monitor
Accurately measure remaining eluent volume using a Thermo Scientific™ Dionex™ Eluent Monitor
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
22185-627072 L Eluent Bottles
22185-627084 L Eluent Bottles
Catalog number 22185-62707
Price (USD)
695.00
Each
Add to cart
For Use With (Equipment):
2 L Eluent Bottles
Price (USD)
695.00
Each
Add to cart
Enable your Thermo Scientific™ Dionex™ ion chromatography (IC) system to accurately measure remaining eluent volumes using the Thermo Scientific™ Dionex™ Eluent Monitors. Dionex Eluent Monitors help operators ensure that there is sufficient eluent for the analyses scheduled to be run, optimizing system uptime and throughput.
Optional accessory for all Dionex IC systems
Specifications
DescriptionDionex Eluent Monitor for 2 L bottles
For Use With (Equipment)2 L Eluent Bottles
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link