Thermo Scientific Chemicals

Bromoacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: 221861000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Bromoacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: 221861000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 221861000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
590-17-0
IUPAC Name
2-bromoacetonitrile
Molecular Formula
C2H2BrN
InChI Key
REXUYBKPWIPONM-UHFFFAOYSA-N
SMILES
BrCC#N
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Refractive index
1.4780 to 1.4810 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text