Thermo Scientific Chemicals

Bromoacetaldehyde dimethyl acetal, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 221870250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Bromoacetaldehyde dimethyl acetal, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 221870250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 221870250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7252-83-7
IUPAC Name
2-bromo-1,1-dimethoxyethane
Molecular Formula
C4H9BrO2
InChI Key
FUSFWUFSEJXMRQ-UHFFFAOYSA-N
SMILES
COC(CBr)OC
GC
>=96.0 %
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Stabilizer
0.2 % potassium carbonate
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text