Bromoacetaldehyde dimethyl acetal, 97%, stabilized, Thermo Scientific Chemicals
Bromoacetaldehyde dimethyl acetal, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bromoacetaldehyde dimethyl acetal, 97%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number 221871000
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
134.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
134.00 
Save 20.35 (15%)
Add to cart
Bromoacetaldehyde dimethyl acetal, 97%, stabilized, Thermo Scientific Chemicals
Catalog number221871000
Price (USD)/ Each
113.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
134.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-bromo-1,1-dimethoxyethane
Molecular FormulaC4H9BrO2
InChI KeyFUSFWUFSEJXMRQ-UHFFFAOYSA-N
SMILESCOC(CBr)OC
Molecular Weight (g/mol)169.02
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Stabilizer0.2 % potassium carbonate
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only