2-Bromothiophenol, 97%, Thermo Scientific Chemicals
2-Bromothiophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromothiophenol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
10 g
Packaging:
Glass bottle
Catalog number 221980500
Price (USD)/ Each
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6320-02-1
IUPAC Name2-bromobenzene-1-thiol
Molecular FormulaC6H5BrS
InChI KeyYUQUNWNSQDULTI-UHFFFAOYSA-N
SMILESSC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.6340 to 1.6370 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only