Search Thermo Fisher Scientific
Description | Resolution Standard; Verification Standard |
Type | Toluene in Hexane Solution Standard |
Wavelength Range | 265 to 270 nm |
For Use With (Equipment) | Evolution 60S, 201, 220 260 Bio, 300, GENESYS 10S, BioMate 3S |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | For Use With (Equipment) | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
222-226600 | Each | Evolution 60S, 201, 220 260 Bio, 300, GENESYS 10S, BioMate 3S | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|