Search Thermo Fisher Scientific
The Thermo Scientific™ Rapid Mixing Accessory, when used with the Evolution Pro UV-Visible Spectrophotometer, provides reliable and accurate measurement of millisecond timescale kinetics, allowing scientists to make kinetic measurements nearly 1000 times faster than allowed by manual reagent mixing.
Description | Rapid Mixing Accessory without Pneumatic Drive |
For Use With (Equipment) | Evolution Pro UV-Visible Spectrophotometer |
Includes | Without Pneumatic Drive |
Unit Size | Each |
Description | Rapid Mixing Accessory with Pneumatic Drive |
For Use With (Equipment) | Evolution Pro UV-Visible Spectrophotometer |
Includes | Pneumatic Drive |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Includes | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
222-234300 | Each | Without Pneumatic Drive | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
222-234400 | Each | Pneumatic Drive | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Reliable, Millisecond Date Collection Rate
Precise Reaction Start Time
Complete Temperature Control
Includes:
Thermocouple and alarm
Compatible with: