2-Propanesulfonyl chloride, 97%, Thermo Scientific Chemicals
2-Propanesulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Propanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 222030250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10147-37-2
IUPAC Namepropane-2-sulfonyl chloride
Molecular FormulaC3H7ClO2S
InChI KeyDRINJBFRTLBHNF-UHFFFAOYSA-N
SMILESCC(C)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow-green
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4530 to 1.4580 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only