Sulfamic acid, 99%, Thermo Scientific Chemicals
Sulfamic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfamic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
250 g
1 kg
2.5 kg
Packaging:
View all
Plastic Drum
Catalog number 222070100
Price (USD)/ Each
427.65
Online exclusive
475.00 
Save 47.35 (10%)
-
Add to cart
Quantity:
10 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)/ Each
427.65
Online exclusive
475.00 
Save 47.35 (10%)
Add to cart
Sulfamic acid, 99%, Thermo Scientific Chemicals
Catalog number222070100
Price (USD)/ Each
427.65
Online exclusive
475.00 
Save 47.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5329-14-6
IUPAC Namesulfamic acid
Molecular FormulaH3NO3S
InChI KeyIIACRCGMVDHOTQ-UHFFFAOYSA-N
SMILESNS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point200°C to 208°C
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only