Chlorobenzene, 99+%, for spectroscopy, Thermo Scientific Chemicals
Chlorobenzene, 99+%, for spectroscopy, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chlorobenzene, 99+%, for spectroscopy, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number 222115000
Price (USD)/ Each
-
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS108-90-7
IUPAC Namechlorobenzene
Molecular FormulaC6H5Cl
InChI KeyMVPPADPHJFYWMZ-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
UVat 288 nm A: =<1.0
UVat 300 nm A: =<0.07
Residue after evaporation=<0.0005 %
UVat 290 nm A: =<0.40
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only