Thermo Scientific Chemicals

Chloroiodomethane, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 222151000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Chloroiodomethane, 98%, stabilized, Thermo Scientific Chemicals

Catalog number: 222151000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 222151000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
593-71-5
IUPAC Name
chloro(iodo)methane
Molecular Formula
CH2ClI
InChI Key
PJGJQVRXEUVAFT-UHFFFAOYSA-N
SMILES
ClCI
Appearance (Color)
Clear colorless to yellow to pink
Infrared spectrum
Conforms
Appearance
May darken on storage
Stabilizer
Copper
Refractive index
1.5810 to 1.5840 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text