Thermo Scientific Chemicals

4-Bromo-N,N-dimethylaniline, 99%, Thermo Scientific Chemicals

Catalog number: 222211000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

4-Bromo-N,N-dimethylaniline, 99%, Thermo Scientific Chemicals

Catalog number: 222211000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 222211000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
586-77-6
IUPAC Name
4-bromo-N,N-dimethylaniline
Molecular Formula
C8H10BrN
InChI Key
XYZWMVYYUIMRIZ-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(Br)C=C1
Titration with HClO4
>=98.5 %
Infrared spectrum
Conforms
Appearance (Color)
White to grey to light blue
Appearance (Form)
Crystalline solid
Melting point
53°C to 56°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text