Poly(sodium-p-styrenesulfonate), average M.W. 70.000
Poly(sodium-p-styrenesulfonate), average M.W. 70.000
Poly(sodium-p-styrenesulfonate), average M.W. 70.000
Poly(sodium-p-styrenesulfonate), average M.W. 70.000
Thermo Scientific Chemicals

Poly(sodium-p-styrenesulfonate), average M.W. 70.000

CAS: 25704-18-1 | (C8H7NaO3S)n | 184.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 222271000
Price (USD)
88.65
Online Exclusive
98.00
Save 9.35 (10%)
Each
In stock
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
88.65
Online Exclusive
98.00
Save 9.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS25704-18-1
Molecular Formula(C8H7NaO3S)n
SMILES[Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-*
MDL NumberMFCD00084449
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link