Thermo Scientific Chemicals

Barium sulfate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 222510010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Barium sulfate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 222510010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 222510010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7727-43-7
IUPAC Name
barium(2+) sulfate
Molecular Formula
BaO4S
InChI Key
TZCXTZWJZNENPQ-UHFFFAOYSA-L
SMILES
[Ba++].[O-]S([O-])(=O)=O
Appearance (Color)
White
Appearance (Form)
Powder
Loss on ignition
=<2.0 % (600°C)
Heavy metals (as Pb)
=<10 ppm
Acidity/alkalinity
Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text