Thermo Scientific Chemicals

Chromium potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 222521000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Chromium potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals

Catalog number: 222521000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 222521000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7788-99-0
IUPAC Name
chromium(3+) potassium dodecahydrate disulfate
Molecular Formula
CrH24KO20S2
InChI Key
ZFVHBEKVAITXHW-UHFFFAOYSA-J
SMILES
O.O.O.O.O.O.O.O.O.O.O.O.[K+].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Appearance (Color)
Violet to dark purple
Appearance (Form)
Crystalline powder or crystals
Titration Iodometric
98.0 to 102.0 %
Chloride (Cl)
=<20 ppm
Ammonium (NH4)
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text