Potassium bromide, 99+%, for analysis, Thermo Scientific Chemicals
Potassium bromide, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium bromide, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 222550025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namepotassium bromide
Molecular FormulaBrK
InChI KeyIOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES[K+].[Br-]
Molecular Weight (g/mol)119.00
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Heavy metals (as Pb)=<5 ppm
Chloride (Cl)=<0.2 %
Sulfate (SO4)=<0.005 %
Barium (Ba)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only