Potassium bromide, 99+%, for analysis, Thermo Scientific Chemicals
Potassium bromide, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium bromide, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 222555000
Price (USD)/ Each
81.65
Online exclusive
90.40 
Save 8.75 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
81.65
Online exclusive
90.40 
Save 8.75 (10%)
Add to cart
Potassium bromide, 99+%, for analysis, Thermo Scientific Chemicals
Catalog number222555000
Price (USD)/ Each
81.65
Online exclusive
90.40 
Save 8.75 (10%)
-
Add to cart
Chemical Identifiers
CAS7758-02-3
IUPAC Namepotassium bromide
Molecular FormulaBrK
InChI KeyIOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES[K+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Bromate (BrO3)=<0.001 %
Appearance (Color)White
Heavy metals (as Pb)=<5 ppm
Chloride (Cl)=<0.2 %
Sulfate (SO4)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only