Tin, granular, 0.1-0.8mm, Thermo Scientific Chemicals
Tin, granular, 0.1-0.8mm, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin, granular, 0.1-0.8mm, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 222571000
Price (USD)/ Each
45.65
Online exclusive
50.90 
Save 5.25 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
45.65
Online exclusive
50.90 
Save 5.25 (10%)
Add to cart
Tin, granular, 0.1-0.8mm, Thermo Scientific Chemicals
Catalog number222571000
Price (USD)/ Each
45.65
Online exclusive
50.90 
Save 5.25 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
Appearance (Form)Granular powder
Antimony (Sb)=<200 ppm
Iron (Fe)=<500 ppm
Bismuth (Bi)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only