Tin, granular, 0.1-0.8mm, Thermo Scientific Chemicals
Tin, granular, 0.1-0.8mm, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin, granular, 0.1-0.8mm, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number 222575000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
Molecular Weight (g/mol)118.71
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
Appearance (Form)Granular powder
Lead (Pb)=<700 ppm
Copper (Cu)=<100 ppm
Particle size0.1 to 0.8 mm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only