Tellurium(IV) oxide, 99+%, Thermo Scientific Chemicals
Tellurium(IV) oxide, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tellurium(IV) oxide, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number 222592500
Price (USD)/ Each
439.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
517.00 
Save 77.35 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
439.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
517.00 
Save 77.35 (15%)
Add to cart
Tellurium(IV) oxide, 99+%, Thermo Scientific Chemicals
Catalog number222592500
Price (USD)/ Each
439.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
517.00 
Save 77.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name(oxo-λ⁴-tellanylidene)oxidane
Molecular FormulaO2Te
InChI KeyLAJZODKXOMJMPK-UHFFFAOYSA-N
SMILESO=[Te]=O
Molecular Weight (g/mol)159.60
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Assay>=99.0 % (TeO2)
Lead (Pb)=<50 ppm
Iron (Fe)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only