Zinc, granular, 30 mesh, Thermo Scientific Chemicals
Zinc, granular, 30 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc, granular, 30 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 222615000
Price (USD)/ Each
93.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
110.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
93.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
110.00 
Save 16.35 (15%)
Add to cart
Zinc, granular, 30 mesh, Thermo Scientific Chemicals
Catalog number222615000
Price (USD)/ Each
93.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
110.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver grey
Appearance (Form)Granules or filings
Titration Complexometric>=99.8 %
Iron (Fe)=<0.01 %
Lead (Pb)=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only