Thermo Scientific Chemicals

Lead, +99%, granular, Thermo Scientific Chemicals

Catalog number: 222625000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Lead, +99%, granular, Thermo Scientific Chemicals

Catalog number: 222625000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 222625000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7439-92-1
IUPAC Name
lead
Molecular Formula
Pb
InChI Key
WABPQHHGFIMREM-UHFFFAOYSA-N
SMILES
[Pb]
Appearance (Form)
Granules
Assay
>=99.0 %
Appearance (Color)
Grey

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text