Thermo Scientific Chemicals

Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf, Thermo Scientific Chemicals

Catalog number: 222635000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Zirconyl(IV) nitrate hydrate, 99.5%, contains up to 4% Hf, Thermo Scientific Chemicals

Catalog number: 222635000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 222635000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14985-18-3
IUPAC Name
(nitrooxy)(oxo)zirconio nitrate
Molecular Formula
N2O7Zr
InChI Key
JWBWFTANBHYHBC-UHFFFAOYSA-N
SMILES
[O-][N+](=O)O[Zr](=O)O[N+]([O-])=O
Gravimetry
calculated on the actual degree of hydration
Gravimetry
calculated on the actual degree of hydration
Degree of hydration
6 to 7
Appearance (Color)
White
Gravimetry
>=99.4 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text