Tin(II) chloride dihydrate, 98+%, Thermo Scientific Chemicals
Tin(II) chloride dihydrate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(II) chloride dihydrate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 222660025
Price (USD)/ Each
537.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
632.00 
Save 94.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
537.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
632.00 
Save 94.35 (15%)
Add to cart
Tin(II) chloride dihydrate, 98+%, Thermo Scientific Chemicals
Catalog number222660025
Price (USD)/ Each
537.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
632.00 
Save 94.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10025-69-1
IUPAC Nameλ²-tin(2+) dihydrate dichloride
Molecular FormulaCl2H4O2Sn
InChI KeyFWPIDFUJEMBDLS-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Sn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Titration Iodimetric98.0 to 103.0 %
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only