Thermo Scientific Chemicals

L(+)-Potassium hydrogen tartrate, 99%, Thermo Scientific Chemicals

Catalog number: 222672500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

L(+)-Potassium hydrogen tartrate, 99%, Thermo Scientific Chemicals

Catalog number: 222672500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 222672500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
868-14-4
IUPAC Name
potassium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate
Molecular Formula
C4H5KO6
InChI Key
KYKNRZGSIGMXFH-ZVGUSBNCSA-M
SMILES
[K+].O[C@H]([C@@H](O)C([O-])=O)C(O)=O
Appearance (Color)
Colorless to white
Loss on drying
=<0.5 % (105°C)
Specific optical rotation
on dried substance
Sulfate (SO4)
=<500 ppm
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text