Thermo Scientific Chemicals

Barium acetate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 222800010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Barium acetate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 222800010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 222800010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
543-80-6
IUPAC Name
barium(2+) diacetate
Molecular Formula
C4H6BaO4
InChI Key
ITHZDDVSAWDQPZ-UHFFFAOYSA-L
SMILES
[Ba++].CC([O-])=O.CC([O-])=O
Nitrate (NO3)
=<20 ppm
Magnesium (Mg)
=<10 ppm
Calcium (Ca)
=<50 ppm
Appearance (Form)
Crystalline powder
pH
6.5 to 8.5 (5 % at 20°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text