Thermo Scientific Chemicals

Nickel(II) acetate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 223145000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Nickel(II) acetate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 223145000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 223145000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6018-89-9
IUPAC Name
nickel(2+) diacetate tetrahydrate
Molecular Formula
C4H14NiO8
InChI Key
OINIXPNQKAZCRL-UHFFFAOYSA-L
SMILES
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O
Titration Complexometric
>=99.0 %
Aluminium (Al)
=<5 ppm
Calcium (Ca)
=<100 ppm
Cadmium (Cd)
=<50 ppm
Cobalt (Co)
=<2000 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text