Thermo Scientific™

Sulfo-SMPB (sulfosuccinimidyl 4-(N-maleimidophenyl)butyrate)

Catalog number: 22317
Have Questions?
Thermo Scientific™

Sulfo-SMPB (sulfosuccinimidyl 4-(N-maleimidophenyl)butyrate)

Catalog number: 22317
Have Questions?
Catalog Number
22317
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2231750 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Sulfo-SMPB is a water-soluble amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length aromatic spacer arm (11.6 angstroms).

Features of Sulfo-SMPB:

Reactive groups: sulfo-NHS ester and maleimide
Reactive towards: amino and sulfhydryl groups
• Extended chain length limits steric hindrance
• Non-cleavable
• Water-soluble (compare to SMPB)
• Membrane impermeable, allowing for cell surface labeling

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
SMPB (succinimidyl 4-(p-maleimidophenyl)butyrate)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Amine-Sulfhydryl
Cleavable
No
Description
Sulfo-SMPB
Molecular Weight (g/mol)
458.37
PEGylated
No
Spacer Arm Length
11.6 Å
Cell Permeability
No
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Heterobifunctional
Reactive Moiety
Maleimide, Sulfo-NHS Ester
Spacer
Medium (10 to 30 Å)
Form
Powder
Quantity
50 mg
Solubility
Water
Format
Standard
Water Soluble
Yes
Product Type
Crosslinker

Contents & Storage

Upon receipt store desiccated at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text