Thermo Scientific Chemicals

N-(2-Aminoethyl)acetamide, 90%, Thermo Scientific Chemicals

Catalog number: 223170050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

N-(2-Aminoethyl)acetamide, 90%, Thermo Scientific Chemicals

Catalog number: 223170050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 223170050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1001-53-2
IUPAC Name
N-(2-aminoethyl)acetamide
Molecular Formula
C4H10N2O
InChI Key
DAKZISABEDGGSV-UHFFFAOYSA-N
SMILES
CC(=O)NCCN
Melting point
45°C to 50°C
Appearance (Form)
Crystalline solid
GC
>=88 %
Appearance (Color)
White to yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text