Thermo Scientific Chemicals

Titanium(IV) n-butoxide, 98+%, Thermo Scientific Chemicals

Catalog number: 223190100
10 kg, Plastic Drum, Each
Thermo Scientific Chemicals

Titanium(IV) n-butoxide, 98+%, Thermo Scientific Chemicals

Catalog number: 223190100
10 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 223190100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5593-70-4
IUPAC Name
titanium(4+) tetrakis(butan-1-olate)
Molecular Formula
C16H36O4Ti
InChI Key
YHWCPXVTRSHPNY-UHFFFAOYSA-N
SMILES
[Ti+4].CCCC[O-].CCCC[O-].CCCC[O-].CCCC[O-]
Appearance (Color)
Clear colorless to yellow to light orange
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Titanium (Ti)
13.8 to 14.4 %
Viscosity
50 to 90 cP (at 25°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text