Thermo Scientific Chemicals

Sodium perborate tetrahydrate, 97%, pure, Thermo Scientific Chemicals

Catalog number: 223200010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium perborate tetrahydrate, 97%, pure, Thermo Scientific Chemicals

Catalog number: 223200010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 223200010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10486-00-7
IUPAC Name
disodium tetrahydroxy-1,2,4,5,3,6-tetraoxadiborinane-3,6-diuide hexahydrate
Molecular Formula
B2H16Na2O14
InChI Key
MCYPYXIRGFEFGL-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Na+].[Na+].O[B-]1(O)OO[B-](O)(O)OO1
pH
10.1 to 10.6 (1 % solution)
Appearance (Form)
Crystalline powder
Appearance (Color)
White
Titration with KMnO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text