1x1 image pixel for data collection
Magnesium chloride, pure
Magnesium chloride, pure
Magnesium chloride, pure
Magnesium chloride, pure
Thermo Scientific Chemicals

Magnesium chloride, pure

CAS: 7786-30-3 | Cl2Mg | 95.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 223210025
Price (USD)
128.65
Online Exclusive
143.00
Save 14.35 (10%)
Each
In stock
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
128.65
Online Exclusive
143.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7786-30-3
IUPAC Namemagnesium(2+) dichloride
Molecular FormulaCl2Mg
InChI KeyTWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES[Mg++].[Cl-].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay contain grey to black particles
Appearance (Color)White
Water=<3 % (K.F.)
Appearance (Form)Powder
Titration Complexometric>=98.0 % (on dry substance)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0450059Certificate of AnalysisMay 02, 2023223210010, 223211000, 223210025, 223210250
A0402852Certificate of AnalysisDec 16, 2022223210010, 223211000, 223210025, 223210250
A0400128Certificate of AnalysisDec 15, 2022223210010, 223211000, 223210025, 223210250
A0395558Certificate of AnalysisDec 14, 2022223210010, 223211000, 223210025, 223210250
A0410218Certificate of AnalysisDec 13, 2022223210010, 223211000, 223210025, 223210250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection