Thermo Scientific Chemicals

Magnesium chloride, pure, Thermo Scientific Chemicals

Catalog number: 223210250
25 kg, Fibre drum, Each
Thermo Scientific Chemicals

Magnesium chloride, pure, Thermo Scientific Chemicals

Catalog number: 223210250
25 kg, Fibre drum, Each
Quantity
Packaging
Catalog number: 223210250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7786-30-3
IUPAC Name
magnesium(2+) dichloride
Molecular Formula
Cl2Mg
InChI Key
TWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES
[Mg++].[Cl-].[Cl-]
Appearance
May contain grey to black particles
Appearance (Color)
White
Water
=<3 % (K.F.)
Appearance (Form)
Powder
Titration Complexometric
>=98.0 % (on dry substance)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text