Sodium chlorite, 80%, pure, unstabilized
Sodium chlorite, 80%, pure, unstabilized
Sodium chlorite, 80%, pure, unstabilized
Sodium chlorite, 80%, pure, unstabilized
Thermo Scientific Chemicals

Sodium chlorite, 80%, pure, unstabilized

Sodium chlorite, 80%, ClNaO2, CAS Number-7758-19-2, 7647-14-5, unii-g538ebv4vf, alcide ld, neo silox d, chlorous acid, sodium salt, caswell no. 755, sodiumchlorite, textone, sodium chlorite, textile, chlorite sodium, 500g, CHEBI:78667, White, 231-836-6, 05,603; 09,423; 11,481; 13,280, 90.44, Pure | CAS: 7758-19-2 | ClNaO2 | 90.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic drum
Catalog number 223230100
Price (USD)
839.00
Each
-
Add to cart
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
839.00
Each
Add to cart
Chemical Identifiers
CAS7758-19-2
IUPAC Namesodium chlorite
Molecular FormulaClNaO2
InChI KeyUKLNMMHNWFDKNT-UHFFFAOYSA-M
SMILES[Na+].[O-][Cl]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodometric>=75 %
Appearance (Color)White
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection